Learn More
Rat IgG1 kappa Isotype Control (eBRG1), PE-Cyanine5, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 15-4301-81
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: PE-Cy5 Rat IgG1 Iso Cntrl has been reported for use in flow cytometric analysis. Applications Tested: This PE-Cy5 Rat IgG1 isotype control antibody should be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: PE-Cy5 Rat IgG1 Iso Cntrl has been reported for use in flow cytometric analysis.Applications Tested: This PE-Cy5 Rat IgG1 isotype control antibody should be used at the same concentration as the experimental antibody.Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
Rat IgG1 kappa | |
eBRG1 | |
PE-Cyanine5 | |
IgG1, IgG1k | |
Affinity chromatography | |
RUO | |
None | |
Liquid |
Flow Cytometry | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Rat | |
50 μg | |
Isotype Control (Primary) | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
IgG1 κ |
For Research Use Only.