Learn More
Rat IgG1 kappa Isotype Control (eBRG1), Biotin, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 13-4301-85
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG1 isotype control can be used in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA. Applications Tested: This rat IgG1 isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and can be used at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: This rat IgG1 isotype control can be used in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA.Applications Tested: This rat IgG1 isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and can be used at the same concentration as the experimental antibody.Filtration: 0.2μm post-manufacturing filtered.Specifications
Rat IgG1 kappa | |
eBRG1 | |
Biotin | |
IgG1, IgG1k | |
Affinity chromatography | |
RUO | |
None | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
0.5mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Rat | |
500 μg | |
Isotype Control (Primary) | |
Store at 2-8°C. Do not freeze. | |
IgG1 κ |
For Research Use Only.