missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG1 kappa Isotype Control (eBRG1), Biotin, eBioscience™, Invitrogen™

Rat Isotype Control

Brand:  Affymetrix eBioscience 13-4301-82

Code : Z2

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13108185

  • £89.25 / 100µg
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG1 isotype control can be used in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA. Applications Tested: This rat IgG1 isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and can be used at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgG1 is a subclass of immunoglobulin G (IgG) antibodies that are commonly used in research and diagnostics. These antibodies are produced by rats in response to an antigen and can be purified from serum or hybridoma cell culture supernatants. Rat IgG1 is often used as a negative control in experiments, as it does not bind to many mammalian antigens. It is also used in immunohistochemistry, flow cytometry, and western blotting to detect specific antigens. Gene targeting is a powerful tool used to manipulate the rat genome and generate animal models for studying human diseases. With gene targeting, specific genes can be knocked out, inserted, or modified to investigate their role in disease pathogenesis.
Specifications

Specifications

Rat IgG1 kappa
eBRG1
Biotin
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
100 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.