missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG VisUCyte HRP Polymer Antibody, R&D Systems™

Goat Polyclonal Antibody
£269.00 - £2180.00
Specifications
| Antigen | IgG |
|---|---|
| Dilution | Immunohistochemistry |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | HRP |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18714278
![]() |
R&D Systems
VC005-025 |
25 mL |
£269.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797818
![]() |
R&D Systems
VC005-050 |
50 mL (2 x 25 mL) |
£433.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18776778
![]() |
R&D Systems
VC005-125 |
125 mL |
£1155.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766739
![]() |
R&D Systems
VC005-250 |
250 mL (2 x 125 mL) |
£2180.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG Polyclonal specifically detects IgG in Rat samples. It is validated for Immunohistochemistry.Specifications
| IgG | |
| Immunohistochemistry | |
| HRP | |
| RUO | |
| Supplied as a solution in PBS with preservative. | |
| 3500 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry | |
| Polyclonal | |
| Goat | |
| Rat | |
| Immunoglobulin G, ImmunoglobulinG | |
| Rat IgG | |
| Secondary | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title