Learn More
R&D Systems™ Rat IFN-gamma ELISpot Development Module, 5 Plate
Product Code. 18421150
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18421150 1 Kit 1 set
1 options
Product Code. 18421150 Supplier R&D Systems™ Supplier No. SEL585
Sufficient for at least five 96-well plates
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.