missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat IFN-beta Recombinant Protein

Product Code. 10414383
Change view
Click to view available options
Quantity:
1 x 10⊃5 U
Unit Size:
Each
Product Code. Quantity unitSize
10414383 1 x 10⊃5 U Each
1 options
This item is not returnable. View return policy

Product Code. 10414383

Brand: Gibco™ PRC4024

This item is not returnable. View return policy

Rat IFN-beta Recombinant Protein

Rat IFN-beta Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IFN-β is involved in immune responses against viral infection.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

For Use With (Application) Cell Culture
Gene ID (Entrez) 24481
Molecular Weight (g/mol) 19.7 kDa
Purification Method Affinity Chromatography
Quantity 1 x 10⊃5 U
Gene Alias IFN-β
Species Rat
Content And Storage Store in refrigerator (2–8°C).
Endotoxin Level 1 EU/vial
Mass 2.4 x 10^5 units
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System Mammalian
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interferons
Research Category Immunology, Virology, Signal Transduction
Product Line Gibco
Purity or Quality Grade 90 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.