missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat IFN-alpha Recombinant Protein
Click to view available options
Quantity:
1 x 10^5 units
Unit Size:
Each
Description
Reconstitution: Dissolve the contents of the vial by injection of 150 μL sterile distilled water. Lyophilized product is stable for a defined length of time at -20°C. After reconstitution, the contents can be best divided into small aliquots for single use and stored at -80°C. After thawing, the cytokine is stable for at least four weeks at 4°C.
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
Specifications
Specifications
| Accession Number | P05011 |
| For Use With (Application) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
| Formulation | Citric acid with 125mM trehalose and no preservative; pH 5 |
| Gene ID (Entrez) | 298210, 298213 |
| Molecular Weight (g/mol) | 19 kDa |
| Name | Rat IFN-alpha |
| pH Range | 5 |
| Purification Method | Purified |
| Quantity | 1 x 10^5 units |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction