missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Rat HSP90 ELISA Kit (Chemiluminescence)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Rat HSP90 ELISA Kit (Chemiluminescence) measures rat HSP90 in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 31.25 to 2000 pg/mL |
| Assay Sensitivity | 18.75 pg/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat HSP-90 in samples. No significant cross-reactivity or interference between Rat HSP-90 and analogues was observed. |
| Gene ID (Entrez) | 3320 |
| Gene Symbol | HSP90AA1 |
| Host Species | Rat |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction