missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Hepassocin/FGL1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1105
This item is not returnable.
View return policy
Description
Hepassocin/FGL1 Polyclonal specifically detects Hepassocin/FGL1 in Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Hepassocin/FGL1 | |
| Polyclonal | |
| Unconjugated | |
| Q5M8C6 | |
| FGL1 | |
| Mouse myeloma cell line NS0-derived recombinant rat Hepassocin Asp25-Val314 Accession # Q5M8C6 | |
| 100 μg | |
| Primary | |
| Detects rat Hepassocin in direct ELISAs and Western blots. | |
| Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2267 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction