missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Growth Hormone Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1566-SP
This item is not returnable.
View return policy
Description
Growth Hormone Monoclonal specifically detects Growth Hormone in Rat samples. It is validated for Western Blot.
Specifications
| Growth Hormone | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P01244 | |
| GH1 | |
| E. coli-derived recombinant rat Growth Hormone Phe27-Phe216 Accession # P01244 | |
| 25 μg | |
| Primary | |
| Detects rat Growth Hormone in direct ELISAs and Western blots. Does not cross-react with recombinant human Growth Hormone. | |
| Rat | |
| Purified |
| Western Blot | |
| 222540 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2688 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction