missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Glut4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 13 publications
Brand: R&D Systems MAB1262-SP
This item is not returnable.
View return policy
Description
Glut4 Monoclonal specifically detects Glut4 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification, Immunocytochemistry.
Specifications
| Glut4 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a 0.2 um filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SLC2A4 | |
| Partially purified vesicles containing Glut4 | |
| 25 μg | |
| Primary | |
| Recognizes an epitope in the cytoplasmic portion of rat Glut4 and has been shown to bind only to the muscle-adipose isoform of the glucose transporter, which is antigenically unique from the other transport proteins (1-3). Recognizes Glut4 in human, monkey, rat, mouse, and rabbit systems. It does not recognize this protein in canine systems. Its ability to bind to glucose transporter from other species has not been tested. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification, Immunocytochemistry | |
| 1F8 | |
| Western Blot 1 μg/mL, Immunohistochemistry 5-25 μg/mL, Immunoprecipitation, Immunoaffinity Purification, Immunocytochemistry 8-25 μg/mL | |
| GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, SLC2A4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6517 | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction