missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat GFR alpha-1/GDNF R alpha-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB560
This item is not returnable.
View return policy
Description
GFR alpha-1/GDNF R alpha-1 Monoclonal specifically detects GFR alpha-1/GDNF R alpha-1 in Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GFR alpha-1/GDNF R alpha-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp313E1012, DKFZp686J0156, GDNF family receptor alpha 1, GDNF family receptor alpha-1, GDNF R alpha-1, GDNF receptor alpha-1, GDNFRAGFR-ALPHA-1, GDNFR-alpha-1, GDNFRMGC23045, GFR alpha1, GFRa-1, GFRA1, GFR-alpha-1, Glial cell line-derived neurotrophic factor receptor alpha, GPI-linked anchor protein, PI-linked cell-surface accessory protein, RET ligand 1, RET1L, RETL1FLJ10561, TGF-beta related neurotrophic factor receptor 1, TGF-beta-related neurotrophic factor receptor 1, TRNR1FLJ31546 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2674 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 81401 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q62997 | |
| GFRA1 | |
| Mouse myeloma cell line NS0-derived recombinant rat GFR alpha-1/GDNF R alpha-1 Asp25-Leu445 Accession # Q62997 | |
| 500 μg | |
| Primary | |
| Detects rat GFR alpha-1/GDNF R alpha-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Ret, recombinant human (rh) GFR alpha-2, rhGFR alpha-3 or rmGFR alpha-2 is observed. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction