missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat GDNF ELISA Kit (Chemiluminescence)

Product Code. 18672041 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18672041 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18672041 Supplier Novus Biologicals™ Supplier No. NBP267992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Chemiluminescence)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Rat GDNF in samples. No significant cross-reactivity or interference between Rat GDNF and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 2668
Gene Symbol GDNF
Quantity 1 Kit
Research Discipline Apoptosis, Biologically Active Proteins, Cancer
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.