missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat FGF-BP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1413-SP
This item is not returnable.
View return policy
Description
FGF-BP Monoclonal specifically detects FGF-BP in Rat samples. It is validated for Western Blot.
Specifications
| FGF-BP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9QY10 | |
| FGFBP1 | |
| Mouse myeloma cell line NS0-derived recombinant rat FGF-BP Glu21-Cys238 Accession # Q9QY10 | |
| 25 μg | |
| Primary | |
| Detects rat FGF-BP in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human FGF-BP is observed. | |
| Rat | |
| Purified |
| Western Blot | |
| 227239 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 17 kDa heparin-binding growth factor-binding protein, FGFBP, FGF-BP, FGFBP1, FGF-BP1, FGFBP-1, FGFBPFGF-binding protein 1, fibroblast growth factor binding protein 1, fibroblast growth factor-binding protein 1, HBp17, HBP1717 kDa HBGF-binding protein, heparin-binding growth factor binding protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9982 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction