missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat Fetal Neural Stem Cell Kit
Rat Fetal Neural Stem Cell Kit
Brand: Gibco™ N7744200
Code : NEW
Additional Details : Weight : 0.50000kg
Description
GIBCO™ Rat Fetal Neural Stem Cells provide superior performance in both adherent cell culture in a matrix and suspended neurosphere cultures. The cells are isolated from the cortex of Sprague-Dawley™ rats at day E14 of gestation. Each vial contains 2 × 106 viable cells. For optimal growth performance, we recommend culturing and expanding the cells in StemPro™ NSC serum-free media.Features of the Gibco™ Rat Fetal Neural Stem Cell Kit include:
About neural stem cells
Neural Stem Cells (NSC) are self-renewing multipotent stem cells of nervous system that can differentiate into neurons, oliogdendrocytes, and astrocytes. Multipotent neural stem cells can be isolated from fetal or adult central nervous system tissue, or derived from embryonic stem cells. In development, neurogenesis takes place in the third to fourth week of gestation and continues in specific regions of the central nervous system. Because of their capacity to generate neurons and glial cells, neural stem cells are a valuable resource, not only for neuroscience, but also for the potential treatment of neurodegenerative disease and neurological disorders. Adult-derived NSCs tend to be regionally restricted and have limited plasticity compared to fetal-derived NSC.
Specifications
• Rat Fetal Neural Stem Cells: store in liquid nitrogen • KnockOut DMEM⁄F-12 Basal Medium: store at 2–8°C, protect from light • StemPro NSC SFM Supplement: store at -5 to -20°C, protect from light • FGF Basic Recombinant Human and EGF Recombinant Human: store at 2–8°C, desiccated |
|
Serum-free | |
Cryopreserved | |
Serum-free |
Neural Stem Cells | |
Mammalian Cell Culture | |
Neural Stem Cell Kit | |
KnockOut™, StemPro™ |
For Research Use Only. Not for use in diagnostic procedures.