missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat EphB1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF542
This item is not returnable.
View return policy
Description
EphB1 Polyclonal specifically detects EphB1 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| EphB1 | |
| Polyclonal | |
| Unconjugated | |
| P09759 | |
| EPHB1 | |
| Mouse myeloma cell line NS0-derived recombinant rat EphB1 Met18-Gln538 Accession # P09759 | |
| 100 μg | |
| Primary | |
| Detects rat EphB1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) EphA3, rmEphA4, rmEphB2, rmEphB3, rmEphB4, and rmEphB6 is observed, less than 1% cross-reactivity with recombinant human (rh) EphA1, rhEphA5, rmEphA7, rmEphA8, and recombinant rat EphA5 is observed. | |
| Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
| Cek6, EC 2.7.10, EC 2.7.10.1, EK6, Elk, EPH receptor B1, eph tyrosine kinase 2, EphB1, EPH-like kinase 6, ephrin type-B receptor 1, EPHT2FLJ37986, hEK6, Net, NETHek6, soluble EPHB1 variant 1, Tyrosine-protein kinase receptor EPH-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2047 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction