missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat Dopamine D1R/DRD1 ELISA Kit (Chemiluminescence)

Product Code. 18114024 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18114024

Brand: Novus Biologicals™ NBP267934

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Rat Dopamine D1R/DRD1 ELISA Kit (Chemiluminescence) measures rat Dopamine D1R/DRD1 in serum, plasma and other biological fluids.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.13-200 ng/mL
Assay Sensitivity 1.88 ng/mL
Cross Reactivity This kit recognizes Rat DRD1 in samples. No significant cross-reactivity or interference between Rat DRD1 and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 1812
Gene Symbol DRD1
Host Species Rat
Quantity 1 Kit
Research Discipline GPCR
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Dopamine D1 R/DRD1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.