missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CXCL7/Thymus Chemokine-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1116-SP
This item is not returnable.
View return policy
Description
CXCL7/Thymus Chemokine-1 Monoclonal specifically detects CXCL7/Thymus Chemokine-1 in Rat samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL7/Thymus Chemokine-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 4-20 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_714943 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5473 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization, ELISA | |
| 186701 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Ppbp | |
| E. coli-derived recombinant rat CXCL7/Thymus Chemokine-1 Ile46-Ile107 Accession # NP_714943 | |
| 25 μg | |
| Primary | |
| Detects rat CXCL7/Thymus Chemokine-1 in ELISAs and Western blots. In sandwich ELISAs, no cross-reactivity or interference with recombinant human CXCL5, recombinant mouse CXCL7/Thymus Chemokine-1, recombinant rat (rr) CXCL1/CINC1, rrCXCL2/CINC3, rrCXCL3/CINC-2 alpha, rrCXCL3/CINC-2 beta, or rrCXCL5 is observed. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction