missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CX3CL1/Fractalkine Chemokine Domain Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB537
This item is not returnable.
View return policy
Description
CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Rat samples. It is validated for Western Blot.
Specifications
| CX3CL1/Fractalkine | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6376 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 96834 | |
| Western Blot 1 ug/mL | |
| O55145 | |
| CX3CL1 | |
| E. coli-derived recombinant rat CX3CL1/Fractalkine Chemokine Domain Gln25-Gly100 Accession # O55145 | |
| 500 μg | |
| Primary | |
| Detects rat CX3CL1/Fractalkine Chemokine in direct ELISAs and Western blots. In ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) CX3CL1 is observed, 10% cross-reactivity with rmCCL11 is observed, and no cross-reactivity with recombinant human (rh) CX3CL1 or rhCCL11 is observed. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction