missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Rat Collagen IV ELISA Kit (Chemiluminescence) measures rat Collagen IV in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 93.75-6000 pg/mL |
| Assay Sensitivity | 56.25 pg/mL |
| Cross Reactivity | This kit recognizes Rat COL4 in samples. No significant cross-reactivity or interference between Rat COL4 and analogues was observed. |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Host Species | Rat |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Collagen IV |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?