missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat CHRM3 (NP_036659) VersaClone cDNA

Product Code. 18755388
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18755388 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18755388 Supplier R&D Systems™ Supplier No. RDC1926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cholinergic Receptor Muscarinic 3

The Rat CHRM3 (NP_036659) VersaClone cDNA clone contains the complete ORF for Muscarinic Acetylcholine Receptor M3/CHRM3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Muscarinic Acetylcholine Receptor M3/CHRM3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Product Type cDNA
Gene rChrm3
Accession Number NP_036659
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1783bp
Gene ID (Entrez) 1131 (Human); 12671 (Mouse); 24260 (Rat)
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Rat
Quantity 10 μg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.