missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat Carboxypeptidase-B Recombinant Protein, PeproTech®

Product Code. 17832383
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 17832383

Brand: Gibco™ 40000500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat Carboxypeptidase-B is a 35.1 kDa protein consisting of 307 amino acids. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19223
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 24271
Molecular Weight (g/mol) 35.1 kDa
Name Rat Carboxypeptidase-B
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; carboxypeptidase B; carboxypeptidase B1; carboxypeptidase B1 (tissue); Cpb; Cpb1; Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; PCPB; procarboxypeptidase B; protaminase; tissue carboxypeptidase B
Common Name Carboxypeptidase B1
Gene Symbol CPB1
Biological Activity Carboxypeptidase-B sequentially cleaves C terminal K and R residues.
Conjugate Unconjugated
Recombinant Recombinant
Sequence ASGHSYTKYN NWETIEAWIQ QVATDNPDLV TQSVIGTTFE GRNMYVLKIG KTRPNKPAIF IDCGFHAREW ISPAFCQWFV REAVRTYNQE IHMKQLLDEL DFYVLPVVNI DGYVYTWTKD RMWRKTRSTM AGSSCLGVDP NRNFNAGWCE VGASRSPCSE TYCGPAPESE KETKALADFI RNNLSTIKAY LTIHSYSQMM LYPYSYDYKL PENYEELNAL VKGAAKELAT LHGTKYTYGP GATTIYPAAG GSDDWSYDQG IKYSFTFELR DTGFFGFLLP ESQIRQTCEE TMLAVKYIAN YVREHLY
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.