missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Rat BCMA / TNFRSF17 Protein, His Tag

Product Code. 17296060
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17296060 1 mg 1mg
17286060 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17296060 Supplier ACROBiosystems Supplier No. BCAR52H31mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.5 kDa.

Specifications

Molecular Weight (g/mol) 7.5 kDa
Quantity 1 mg
Source Rat BCMA, His Tag (BCA-R52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # D3ZKQ8-1).
Immunogen Met 1 - Thr 49
Storage Requirements -20°C
Gene Alias TNFRSF17,CD269,BCM,BCMA
Common Name BCMA
Conjugate Unconjugated
Species Rat
Protein Tag His Tag
Expression System HEK293
Form Powder
Protein Subtype Bio-Markers & CD Antigens
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.