missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat Bcl-xL ELISA Kit (Colorimetric)

Product Code. 18606430 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18606430 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18606430 Supplier Novus Biologicals™ Supplier No. NBP2770431Kit

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 46.88 pg/mL to 3000 pg/mL
Assay Sensitivity 11.72 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Tissue Homogenates, Cell Lysates
Specificity This assay has high sensitivity and excellent specificity for detection of Rat Bcl-2. No significant cross-reactivity or interference between Rat Bcl-2 and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 598
Gene Symbol BCL2L1
Quantity 1 Kit
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors
Storage Requirements Store at 4°C.
Synonym Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.