missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat B7-2/CD86 Antibody, R&D Systems™

Goat Polyclonal Antibody
£180.00 - £496.00
Specifications
| Antigen | B7-2/CD86 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18716138
![]() |
R&D Systems
AF1340-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18058921
|
R&D Systems
AF1340 |
100 μg |
£496.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-2/CD86 Polyclonal specifically detects B7-2/CD86 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| B7-2/CD86 | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Polyclonal | |
| Goat | |
| Rat | |
| Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
| CD86 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA | |
| Unconjugated | |
| RUO | |
| O35531 | |
| 942 | |
| Mouse myeloma cell line NS0-derived recombinant rat B7-2/CD86 Val29-Ile250 Accession # O35531 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects rat B7-2/CD86 in ELISAs and Western blots. In sandwich immunoassays, less than 20% cross-reactivity with recombinant mouse (rm) B7-2 is observed and less than 0.2% cross-reactivity with recombinant rat B7-1, rmB7-1, and recombinant human B7-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title