missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smc3, Rat, Clone: KT75, Abnova™
Rat monoclonal antibody raised against recombinant Smc3.
Brand: Abnova MAB10224.100ug
This item is not returnable.
View return policy
Specifications
| Smc3 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Smc3 | |
| Smc3 | |
| Recombinant GST fusion protein corresponding to mouse Smc3. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
| Western Blot | |
| KT75 | |
| Rat monoclonal antibody raised against recombinant Smc3. | |
| In PBS (0.09% sodium azide) | |
| Bamacan/Cspg6/HCAP/Mmip1/SmcD | |
| Rat | |
| Protein G purification | |
| RUO | |
| 13006 | |
| Store at 4°C for one month. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction