missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHLDA3, Rat, Clone: 4B6, Abnova™
Rat monoclonal antibody raised against synthetic peptide of PHLDA3.
Brand: Abnova MAB6755.100ug
This item is not returnable.
View return policy
Specifications
| PHLDA3 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PHLDA3 | |
| PHLDA3 | |
| A synthetic peptide corresponding to amino acids 1-31 of human PHLDA3. | |
| RUO | |
| 23612 | |
| Human | |
| Liquid |
| Western Blot | |
| 4B6 | |
| Rat monoclonal antibody raised against synthetic peptide of PHLDA3. | |
| In PBS (50% glycerol) | |
| TIH1 | |
| Rat | |
| 100 μg | |
| Primary | |
| This antibody is specific to human PHLDA3. | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction