missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgM Heavy Chain Secondary Antibody, Biotin, Novus Biologicals™
Rat Monoclonal Secondary Antibody
Brand: Novus Biologicals NBP1-43527-0.1MG
This item is not returnable.
View return policy
Description
IgM Heavy Chain Monoclonal specifically detects IgM Heavy Chain in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IgM | |
| Monoclonal | |
| Biotin | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
| IGHM | |
| The immunogen for this antibody was IgM. | |
| 0.1 mg | |
| Immunology | |
| 3507 | |
| Store at 4C. Do not freeze. |
| Flow Cytometry | |
| eB121-15F9 | |
| Flow Cytometry | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction