missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgG2b Heavy Chain Secondary Antibody (LO-MG2b-2), PE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG2b Heavy Chain Monoclonal antibody specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IgG2b Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LO-MG2b-2 |
| Conjugate | PE |
| Formulation | PBS |
| Host Species | Rat |
| Immunogen | Purified mouse IgG2b |
| Purification Method | Protein A purified |
| Quantity | 0.25 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?