missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgG2a Secondary Antibody, Invitrogen™
Rat Monoclonal Secondary Antibody
Brand: Invitrogen SA135642
Code : LD
Additional Details : Weight : 0.01000kg
Description
Mouse IgG2a Monoclonal antibody specifically detects Mouse IgG2a in Mouse samples. It is validated for ELISA, Immunohistochemistry (Frozen).Specifications
Mouse IgG2a | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Immunoglobulins. | |
1 mg | |
Secondary | |
4° C | |
IgG2a |
ELISA, Immunohistochemistry (Frozen) | |
LO-MG2a-2 | |
Unconjugated | |
Rat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |