Learn More
Rat anti-Mouse IgG2a, Clone: R19-15, AKP, Secondary Antibody, BD
Brand: BD Biosciences 553389
Description
The R19-15 antibody recognizes an epitope in the CH3 domain of mouse IgG2a, with strong reactivity to the Igh-I[a] allotype and weaker reactivity to Igh-I[b]. It does not react with other Ig isotypes. Molecular genetic analyses suggest that the Igh-I[b] a
Host Species: Rat
Clone: R19-15
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled BALB/c and C57BL/6 mouse Ig
ELISA
Specifications
| Rat anti-Mouse IgG2a | |
| R19-15 | |
| Rat | |
| RUO | |
| Secondary | |
| IgG1 κ |
| Monoclonal | |
| Alkaline Phosphatase (AP) | |
| 1 mL | |
| Immunology | |
| Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.