Learn More
Invitrogen™ Rat anti-Mouse IgG1 Secondary Antibody, Super Bright™ 436, eBioscience™
Rat Monoclonal Secondary Antibody
Brand: Invitrogen™ 62-4015-82
Description
Description: The monoclonal antibody M1-14D12 recognizes mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies. Applications Reported: This M1-14D12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This M1-14D12 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are...
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgG1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry | |
| M1-14D12 | |
| Super Bright 436 | |
| Rat | |
| 100 μg | |
| Secondary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.