Learn More
Rat anti-Mouse IgG1 Secondary Antibody, HRP, eBioscience™, Invitrogen™
Rat Monoclonal Secondary Antibody
£80.25 - £183.00
Specifications
Antigen | Mouse IgG1 |
---|---|
Clone | M1-14D12 |
Concentration | 0.5 mg/mL |
Applications | ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15508306
|
Affymetrix eBioscience
18-4015-80 |
25 μg |
£80.25
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518306
|
Affymetrix eBioscience
18-4015-82 |
100 μg |
£183.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies. Applications Reported: This M1-14D12 antibody has been reported for use in western blotting, ELISA, immunohistochemical and immunocytochemical staining. Applications Tested: This M1-14D12 antibody has been tested by immunocytochemistry and immunohistochemistry after staining with a primary antibody of the mouse IgG1 isotype. This can be used at less than or equal to 20 µg/mL. For western blotting, this can be used at 1 µg/mL (1/500 dilution). It is recommended that the antibody be titrated for optimal performance in the assay of interest.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgG1 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and no preservative; pH 7.2 | |
Secondary | |
4° C |
M1-14D12 | |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
HRP | |
Rat | |
Mouse | |
IgG | |
Affinity chromatography |
For Research Use Only.