missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgA Heavy Chain Secondary Antibody (LO-MA-10), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-68446-250UG
This item is not returnable.
View return policy
Description
IgA Heavy Chain Monoclonal specifically detects IgA Heavy Chain in Mouse samples. It is validated for ELISA.
Specifications
| IgA Heavy Chain | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA | |
| Rat | |
| Affinity Purified | |
| Rat anti Mouse IgA Heavy Chain antibody, clone LO-MA-10, recognizes the alpha heavy chain of mouse immunoglobulin. It does not react with human IgG or IgM. It also does not react with chicken, rabbit, goat, sheep, bovine, horse, dog, pig or baboon IgG. | |
| Store at 4°C. |
| ELISA | |
| LO-MA-10 | |
| Unconjugated | |
| PBS | |
| Mouse IgA | |
| 250 μg | |
| Mouse | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction