Learn More
CD16/CD32 Rat anti-Mouse, Brilliant Violet 510, Clone: 2.4G2, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 740111
Description
The 2.4G2 antibody specifically recognizes a common nonpolymorphic epitope on the extracellular domains of the mouse Fc γIII (CD16) and Fc γII (CD32) Receptors. It has also been reported to bind the Fc γI receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the Fc γIII and Fc γII, and possibly Fc γI, Receptors . CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
| CD16/CD32 | |
| Monoclonal | |
| 0.2mg/mL | |
| FcγRIII/FcγRII; Fcgr3/Fcgr2 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| 2.4G2 | |
| Brilliant Violet 510 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse BALB/c Macrophage J774 | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only