Learn More
CD134 Rat anti-Mouse, Brilliant Violet 650, Clone: OX-86, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 740545
Description
The OX-86 monoclonal antibody specifically binds to the OX-40 antigen (CD134), also known as OX-40L receptor. CD134 is a 50-kDa type-I membrane glycoprotein that belongs to the NGFR/TNFR superfamily. Mouse CD134 is expressed on activated CD4+ and CD8+ T lymphocytes and has been shown to be the sole receptor for the OX-40 Ligand (OX-40L). In the brains of mice with actively induced experimental allergic encephalomyelitis, the expression of CD134 on CD4+ T lymphocytes correlates with disease progression. The OX-40/OX-40L system supplies a costimulatory signal for T-cell proliferation and B-cell proliferation and differentiation. In addition, OX-40 antigen provides a costimulatory signal that induces T cells to proliferate in a CD28-independent manner. In the intact animal, CD134 does not appear to be essential for many T-cell responses, but it seems to play a major role in the pathogenesis of some autoimmune diseases. The OX-86 mAb stains both CD4+ and CD8+ activated T cells, and this expression pattern has been confirmed using OX-40L-Ig fusion protein. CD134 was also detected, using OX-86 mAb, on B cells after stimulation with anti-IgM plus anti-CD40 mAb HM40-3 (Cat. no. 553721). OX-86 mAb does not block binding of OX-40L to OX-40, and it stimulates T-cell proliferation mildly.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
| CD134 | |
| Monoclonal | |
| 0.2mg/mL | |
| Tnfrsf4; Ox40; OX40L receptor; Ly-70; ACT35; Txgp1; TXGP1L | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| OX-86 | |
| Brilliant Violet 650 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Mouse OX-40 3/4 CD4 Chimeric Protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only