Learn More
LPAR2, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LPAR2.
Brand: Abnova PAB24995.100uL
Description
This gene encodes a member of family I of the G protein-coupled receptors, as well as the EDG family of proteins. This protein functions as a lysophosphatidic acid (LPA) receptor and contributes to Ca2+ mobilization, a critical cellular response to LPA in cells, through association with Gi and Gq proteins. An alternative splice variant has been described but its full length sequence has not been determined. [provided by RefSeq]
Specifications
| LPAR2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| LPAR2 | |
| LPAR2 | |
| A synthetic peptide corresponding to LPAR2. | |
| 100 μL | |
| Primary | |
| LPAR2 polyclonal antibody detects endogenous levels of LPAR2 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of LPAR2. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| EDG-4/EDG4/FLJ93869/LPA2 | |
| Rat | |
| Antigen affinity purification | |
| RUO | |
| 9170 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only