missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klrc1/Klrc2/Klrc3, Rat, Biotin, Clone: 20d5, Abnova™
Rat monoclonal antibody raised against Klrc1/Klrc2/Klrc3.
Brand: Abnova MAB5773.500ug
This item is not returnable.
View return policy
Specifications
| Klrc1/Klrc2/Klrc3 | |
| Monoclonal | |
| Biotin | |
| Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| CD159a/NKG2A/NKG2B | |
| Rat | |
| 500 μg | |
| Primary | |
| Mouse NKG2 (Mr ∼ 43 KDa). | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
| Flow Cytometry | |
| 20d5 | |
| Rat monoclonal antibody raised against Klrc1/Klrc2/Klrc3. | |
| In PBS (0.09% sodium azide) | |
| Klrc1 | |
| CHO cells transfected with murine NKG2. | |
| RUO | |
| 16641/16642/58179 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction