missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL27, Rat, Clone: 3H12.F10, Abnova™
Rat monoclonal antibody raised against recombinant Il27.
Brand: Abnova MAB10274.100ug
This item is not returnable.
View return policy
Specifications
| IL27 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Il27 | |
| Il27 | |
| Recombinant protein corresponding to mouse Il27. | |
| RUO | |
| 246779 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 3H12.F10 | |
| Rat monoclonal antibody raised against recombinant Il27. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| IL-27/IL-27p28/Il30/p28 | |
| Rat | |
| 100 μg | |
| Primary | |
| This antibody is specific for mouse and rat p28 protein. Cross-reactivity with IL-27 from other sources has not been determined. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction