missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F2, Rat, Monoclonal Antibody, Abnova™
Rat monoclonal antibody raised against native F2.
Brand: Abnova MAB0285.100ug
This item is not returnable.
View return policy
Specifications
| F2 | |
| Monoclonal | |
| Rat monoclonal antibody raised against native F2. | |
| In PBS (50% glycerol) | |
| Cf-2/Cf2/FII | |
| Rat | |
| 100 μg | |
| Primary | |
| This antibody recognizes mouse prothrombin. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| F2 | |
| F2 | |
| Native purified Mouse F2. | |
| RUO | |
| 14061 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction