missing translation for 'onlineSavingsMsg'
Learn More

Dinitrophenyl Hapten, Rat, Clone: LO-DNP-2, Abnova™

Product Code. 16096726
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16096726 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16096726 Supplier Abnova Supplier No. MAB5195.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat monoclonal antibody raised against Dinitrophenyl Hapten.

Specifications

Antigen Dinitrophenyl Hapten
Applications ELISA
Classification Monoclonal
Clone LO-DNP-2
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Dinitrophenyl Hapten.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Rat
Immunogen DNP-ASCARIS, Nippostrongylus brasiliensis-DNP
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Rat monoclonal antibody against Dinitrophenyl hapten.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ, IgK1a allotype
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.