missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clasp2, Rat, Clone: KT68, Abnova™
Rat monoclonal antibody raised against partial recombinant Clasp2.
Brand: Abnova MAB9738.100ug
This item is not returnable.
View return policy
Specifications
| Clasp2 | |
| Monoclonal | |
| 1 mg/mL | |
| Rat monoclonal antibody raised against partial recombinant Clasp2. | |
| In PBS (0.09% sodium azide) | |
| 1500004F14Rik/8030404L10Rik/C77448/CLASP2beta/mKIAA0627 | |
| Rat | |
| Protein G purification | |
| RUO | |
| 76499 | |
| Store at 4°C for 1 month. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| Immunofluorescence, Western Blot | |
| KT68 | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunofluorescence (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Clasp2 | |
| Clasp2 | |
| Recombinant GST fusion protein corresponding to N-terminus of mouse Clasp2. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction