missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clasp1, Rat, Clone: KT67, Abnova™
Rat monoclonal antibody raised against partial recombinant Clasp1.
Brand: Abnova MAB9737.100ug
This item is not returnable.
View return policy
Specifications
| Clasp1 | |
| Monoclonal | |
| 1 mg/mL | |
| Rat monoclonal antibody raised against partial recombinant Clasp1. | |
| In PBS (0.09% sodium azide) | |
| 1700030C23Rik/5730583A19Rik/B130045P17Rik/KIAA0622/mKIAA0622 | |
| Rat | |
| Protein G purification | |
| RUO | |
| 76707 | |
| Store at 4°C for 1 month. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a |
| Immunofluorescence, Western Blot | |
| KT67 | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunofluorescence (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Clasp1 | |
| Clasp1 | |
| Recombinant GST fusion protein corresponding to N-terminus of mouse Clasp1. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction