missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ahsa1 Rat anti-Human, Mouse, Rat, Clone: 25F2.D9, Abnova™
Rat monoclonal antibody raised against Ahsa1.
Brand: Abnova MAB6668.100ug
This item is not returnable.
View return policy
Specifications
| Ahsa1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Ahsa1 | |
| Ahsa1 | |
| Mouse Aha1. | |
| RUO | |
| 217737 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 25F2.D9 | |
| Rat monoclonal antibody raised against Ahsa1. | |
| In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 | |
| BC023857/MGC36589/MGC36618/p38 | |
| Rat | |
| 100 μg | |
| Primary | |
| Approximately 38 KDa, can run at up to 45kD on SDS-PAGE. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction