Learn More
Ahsa1, Rat, Clone: 25F2.D10, Abnova™
Rat monoclonal antibody raised against Ahsa1.
Brand:  Abnova MAB6669.100ug
Product Code. 16201680
  • £634.00 / 100µg
Specifications
Ahsa1
Monoclonal
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Ahsa1
Ahsa1
Mouse Aha1.
RUO
217737
Human, Mouse, Rat
Liquid
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
25F2.D10
Rat monoclonal antibody raised against Ahsa1.
In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2
BC023857/MGC36589/MGC36618/p38
Rat
100 μg
Primary
Approximately 38 KDa, can run at up to 45 KDa on SDS-PAGE.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG2a κ