Learn More
ABCC10, Rat, Clone: M7I-3, Abnova™
Rat monoclonal antibody raised against partial recombinant ABCC10.
Brand: Abnova MAB6672.1mL
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This ABC full-transporter is a member of the MRP subfamily which is involved in multi-drug resistance. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq
Specifications
| ABCC10 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Frozen sections) (1:20) Immunocytochemistry (1:20-1:50) Wstern Blot (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| ABCC10 | |
| ABCC10 | |
| Recombinant protein corresponding to amino acids 194-272 of human ABCC10. | |
| RUO | |
| 89845 | |
| Human | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
| M7I-3 | |
| Rat monoclonal antibody raised against partial recombinant ABCC10. | |
| In serum-free culture supernatant (0.7% BSA, 0.09% sodium azide) | |
| EST182763/MRP7/SIMRP7 | |
| Rat | |
| 1 mL | |
| Primary | |
| M7I-3 reacts with an internal epitope of MRP7 (ABCC10), an approximately 160 kD transmembrane protein that is related to the multidrug resistance protein MRP1. | |
| Store at 4°C. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.