missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat ALK-7 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5771
This item is not returnable.
View return policy
Description
ALK-7/Activin Receptor Type 1C Monoclonal specifically detects ALK-7/Activin Receptor Type 1C in Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| ALK-7/Activin Receptor Type 1C | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| NP_620790 | |
| ACVR1C | |
| Mouse myeloma cell line NS0-derived recombinant rat ALK-7 Leu26-Arg108 Accession # NP_620790 | |
| 100 μg | |
| Primary | |
| Detects rat ALK-7 in Western blots. In Western blots, no cross-reactivity with recombinant human Activin R1B is observed. | |
| Rat | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 118311 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 130399 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction