missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rat Albumin ELISA Kit (Colorimetric)

Product Code. 18642517 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18642517 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18642517 Supplier Novus Biologicals™ Supplier No. NBP2600941Kit

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.01 μg/mL to 40 μg/mL
Assay Sensitivity 0.01 μg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Urine
Specificity This assay has high sensitivity and excellent specificity for detection of rat MAU. No significant cross-reactivity or interference between rat MAU and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 213
Gene Symbol ALB
Quantity 1 Kit
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Storage Requirements Store at 4°C.
Synonym albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.