missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat AANAT (aa 198-208) Synthetic Peptide
Description
Rat AA-NAT (residues 198-208) Synthetic Peptide, PEP-065, from Invitrogen.
PEP-065 is a 11 amino acid synthetic peptide whose sequence corresponds to residues 198-208 of rat AA-NAT. The sequence of this peptide is (amino to carboxy terminus): C198-H-T-F-L-R-R-N-S-G-C208This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and AA-NAT, catalog # PA1-762. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 25120 |
| Species | Rat |
| Purification Method | purified |
| Accession Number | Q64666 |
| Common Name | Rat AANAT (aa 198-208) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?