missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASSF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101813
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RASSF6.
RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.
Specifications
| RASSF6 | |
| Polyclonal | |
| Unconjugated | |
| Rassf6 | |
| 1600016B17Rik; DKFZp686K23225; putative RAS binding protein; Ras association (RalGDS/AF-6) domain family 6; Ras association (RalGDS/AF-6) domain family member 6; Ras association domain family member 6; ras association domain-containing protein 6; RASSF6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 166824 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6ZTQ3 | |
| Rassf6 | |
| A synthesized peptide derived from human RASSF6(Accession Q6ZTQ3), corresponding to amino acid residues F135-T185. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering